Merck
  • Tris(hydroxymethyl)aminomethane-functionalized agarose particles: parameters affecting the binding of bovine serum albumin.
Tris(hydroxymethyl)aminomethane-functionalized agarose particles: parameters affecting the binding of bovine serum albumin.
Journal of separation science (2012-06-29)
Bin Zhang, Ye Wang, Min Gao, Ming Gu, Changhai Wang
ABSTRACT
A new protein adsorbent is introduced based on the coupling of the common buffer ion, tris(hydroxymethyl)aminomethane, to the agarose gel Sepharose HP from GE Healthcare Bio-Sciences AB, Uppsala, Sweden. The article describes the synthesis of the new adsorbent and the use of BSA as a model in a binding study. By optimization of the coupling procedure, a maximum ligand density of 63.5 μmol/mL gel could be obtained. Adsorption equilibria were investigated in the pH range 5.0-8.0 and at salt concentrations of 0-0.4 mol/L. Binding of BSA under different conditions indicated that both electrostatic interaction and hydrogen bonding were involved in the adsorption process where the former played a major role.
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
SAFC
Tromethamine