Merck
  • Analysis of four alkaloids of Coptis chinensis in rat plasma by high performance liquid chromatography with electrochemical detection.
Analysis of four alkaloids of Coptis chinensis in rat plasma by high performance liquid chromatography with electrochemical detection.
Analytica chimica acta (2012-07-10)
Lihong Liu, Zilin Chen
ABSTRACT
A sensitive and precise high performance liquid chromatography (HPLC)-electrochemical detection (ECD) method has been developed for the simultaneous determination of four isoquinoline alkaloids including berberine, jatrorrhizine, coptisine and palmatine in Chinese medicine Coptis chinensis. The typical HPLC analysis was performed on WondaSil(®) C18-WR column (250×4.6 mm, 5 μm) with the mobile phase comprising 40 mM phosphate buffer (pH 7.0)-acetonitrile (40:60, v/v) at the flow rate of 0.8 mL min(-1). The electrochemical detection employed a three electrode system with a bare glassy carbon electrode at +1.3 V versus the Ag/AgCl reference electrode. The limits of detection (LODs) of four alkaloids ranged from 0.01 to 0.03 μmol L(-1) and the LOD of berberine was 80 times lower than LOD obtained by UV detection. The rat plasma samples were assayed after oral administration of the traditional Chinese medicine Coptis chinensis by the proposed HPLC-ECD method. The recoveries of this method were ranging from 88.0 to 116%, with the relative standard deviation lower than 3.1% for intra-day precision and 5.7% for inter-day precision. These results show that HPLC-ECD is a useful tool for the quality control of herbal medicine Coptis chinensis and also for pharmacokinetic studies.