Merck
  • Studies in search of selective detection of isomeric biogenic hexen-1-ols and hexanal by flowing afterglow tandem mass spectrometry using [H3O]+ and [NO]+ reagent ions.
Studies in search of selective detection of isomeric biogenic hexen-1-ols and hexanal by flowing afterglow tandem mass spectrometry using [H3O]+ and [NO]+ reagent ions.
Rapid communications in mass spectrometry : RCM (2012-07-11)
Frederik Dhooghe, Robbe Vansintjan, Niels Schoon, Crist Amelynck
ABSTRACT
Plants emit a blend of oxygenated volatile C(6) compounds, known as green leaf volatiles (GLVs), in response to leaf tissue damage related to stress conditions. On-line chemical ionization mass spectrometry (CI-MS) techniques have often been used to study the dynamics of these emissions but they fail to selectively detect some important GLV compounds. A flowing afterglow tandem mass spectrometer (FA-TMS) was used to investigate the feasibility of selective on-line detection of isomeric hexen-1-ols and hexanal. Product ions at m/z 101 and 83 from chemical ionization (CI) of these compounds by [H(3)O](+), and product ions at m/z 100, 99, 83, 82 and 72 from CI by [NO](+), have been subjected to collision-induced dissociation (CID) in the collision cell of the TMS at center-of-mass energies ranging between 0 and 9 eV. CID of product ions at m/z 101 and 83 from CI of GLVs with [H(3)O](+) and of product ions at m/z 83, 82 and 72 from CI of GLVs with [NO](+) resulted in identical fragmentation patterns for all measured compounds, ruling out any selectivity. However, CID of product ions at m/z 100 and 99 from CI by [NO](+) led to CID product ions with abundances differing largely between the compounds, allowing the fast selective detection of 2-hexen-1-ols, 3-hexen-1-ols and hexanal with a chosen accuracy within a well-defined range of relative concentrations. This research illustrates that, in contrast to common CI-MS techniques, FA-TMS allows the selective detection of hexanal in a mixture of hexanal and hexen-1-ols with a chosen accuracy for a well-defined range of relative concentrations and represents a step forward in the search for selective detection of GLVs in CI-TMS.
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
cis-3-Hexen-1-ol, 98%
Supelco
cis-3-Hexen-1-ol, analytical standard
Sigma-Aldrich
Hexanal, ≥97%, FCC, stabilized, FG
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
trans-3-Hexen-1-ol, ≥95%, stabilized
Sigma-Aldrich
trans-3-Hexen-1-ol, 97%