Merck
  • Direct rapid analysis of multiple PPCPs in municipal wastewater using ultrahigh performance liquid chromatography-tandem mass spectrometry without SPE pre-concentration.
Direct rapid analysis of multiple PPCPs in municipal wastewater using ultrahigh performance liquid chromatography-tandem mass spectrometry without SPE pre-concentration.
Analytica chimica acta (2012-07-14)
Ke Yu, Bing Li, Tong Zhang
ABSTRACT
Ultrahigh performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) was utilized to develop a rapid, sensitive and reliable method without solid phase extraction (SPE) pre-concentration for trace analysis of 11 pharmaceuticals and personal care products (PPCPs) in influent and effluent from municipal wastewater treatment plants (WWTPs). This method not only shortened the analysis time but also reduced analysis cost significantly by omitting SPE process and avoiding the consumption of SPE cartridge. Detection parameters for UHPLC-MS/MS analysis were optimized, including sample pH, eluent, mobile phase (solvent and additive), column temperature, and flow rate. Under the optimal conditions, all analytes were well separated and detected within 8.0min by UHPLC-MS/MS. The method quantification limits (MQLs) for the 11 PPCPs ranged from 0.040 to 88ngL(-1) and from 0.030 to 90ngL(-1) for influent and effluent, respectively. The matrix effect was systematically investigated and quantified for different types of samples. The analysis of influent and effluent samples of two WWTPs in Hong Kong revealed the presence of 11 PPCPs, including acyclovir, benzophenone-3, benzylparaben, carbamazepine, ethylparaben, fluconazole, fluoxetine, methylparaben, metronidazole, propylparaben, and ranitidine. Their concentrations ranged from 9.1 to 1810ngL(-1) in influent and from 6.5 to 823ngL(-1) in effluent samples collected from Hong Kong WWTPs.
Product Description
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Oxybenzone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
Benzyl 4-hydroxybenzoate, 99%
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Oxybenzone, analytical standard
Ethyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Supelco
Ranitidine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
2-Hydroxy-4-methoxybenzophenone, 98%
Sigma-Aldrich
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
Sigma-Aldrich
Ranitidine hydrochloride, solid