Merck
[Chemical constituents contained in seeds of Notopterygium franchetii].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-07-17)
Yanxia Zhang, Shunyuan Jiang, Kaijie Xu, Haili Shi, Yi Zhou, Wenlong Deng, Lisheng Ding, Shulin Peng
ABSTRACT
To study the chemical constituents from the seeds of Notopterygium franchetii. Ethanol extracts of seeds N. franchetii were separated and purified by such methods as normal and reversed phase column chromatographies and thin-layer chromatography and structurally elucidated by MS and NMR evidences. Twenty nine compounds were separated, they were isoimperatorin (1), [3-sitosterol (2), phellopterin (3), bergapten (4), N-tetra, hexa, octacosanoylanthranilic acid (5-7), daucosterol (8), oxypeucedanin hydrate (9), umbelliferone (10), demethylfuropinnarin (11), (2S, 3S, 4R, 8E)-2-[(2'R)- 2'-hydroxydoco, trico, tetraco, entaco, hexaco sanosylamino] -octadecene-1, 3, 4-triol (12-16), (-)-oxypeucedanin (17), diosmetin (18), bergaptol-O-beta-D-glucopyranoside (19), nodakenin (20), 1'-O-beta-D-glucopyranosyl-(2R, 3S)-3-hydroxynodakenetin (21), uracil (22), decuroside V (23), 8-O-beta-D-glucopyranosyl-5-hydroxypsoralen (24), 8-O-beta-D-glucopyranosyl-5-methoxylpsoralen (25), diosmin (26), alaschanioside C (27), kynurenic acid (28) and mannitol (29). All of these compounds were separated from the seeds of N. franchetii for the first time. Of them, 18, 22, 26 and 29 were firstly obtained from genus Notopterygium.
Sigma-Aldrich
Diosmetin
Supelco
Diosmin, analytical standard
Supelco
β-Sitosterol β-D-glucoside, analytical standard
Sigma-Aldrich
5-Methoxypsoralen, 99%
Supelco
Xanthotoxin, analytical standard
Supelco
8-Methoxypsoralen, analytical standard
Supelco
Bergapten, analytical standard