Merck
  • Separation and determination of chiral composition in penicillamine tablets by capillary electrophoresis in a broad pH range.
Separation and determination of chiral composition in penicillamine tablets by capillary electrophoresis in a broad pH range.
Electrophoresis (2012-07-19)
Lijuan Song, Zhenpeng Guo, Yi Chen
ABSTRACT
A chiral capillary electrophoretic method with nearly full pH window was explored for the separation and determination of dl-penicillamine. A facile one-pot labeling technique was coupled in the method for introduction of chromophore and charge groups onto the analytes to facilitate the electromigration and sensitive detection. By using simply a cost-effective neutral β-cyclodextrin as chiral selector, baseline separation of the dl-penicillamine was achieved from pH 2.0 to over pH 10. Quantification of standard d- and l-penicillamines was demonstrated by taking pH 4.5, 7.4, and 9.7 as the representatives of acidic, neutral, and basic conditions. The working curves were constructed between peak area and concentration, having linear ranges of 8.56-8.56 × 10(2) μg/mL for pH 4.5 and 8.56-1.71 × 10(3) μg/mL for pH 7.4 and 9.7, with correlation coefficients all better than 0.999. The limit of detection (S/N = 3) was 2.58 μg/mL in acidic and neutral conditions or 1.41 μg/mL in basic condition. The method was further validated by assaying the commercial penicillamine tablets, applicable to quantification of the effective enantiomer and the trace impurity of l-penicillamine at a content of down to 0.2, 0.6, and 2.0% for pH 9.7, 4.5, and 7.4, respectively. The recovery determined by spiking technique was in a range from 93.1 to 105 %. The method is easily extendable to the analysis of other chiral amines or amino acids.
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
D-Penicillamine, 98-101%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%