Merck
  • [Effect of opioid peptides on oxygen-dependent microbicidity of peripheral blood neutrophils].
[Effect of opioid peptides on oxygen-dependent microbicidity of peripheral blood neutrophils].
Fiziologiia cheloveka (2012-07-27)
S V Geĭn, K G Gorshkova
ABSTRACT
Investigation ofopioid peptide effect on the production of reactive oxygen species by neutrophils in non-fractionated leukocyte suspension and in purified fraction of peripheral blood neutrophils is disclosed in this work. It was determined that selective delta- and micro-agonists of peptide origin stimulated the spontaneous and suppressed 15 mkg/ml zymosan-induced LDCL (luminol-dependent chemiluminescence) reaction of neutrophils in leukocyte suspension. beta-endorphin was found to render less marked suppressive action on 15 mkg/ml zymosan-induced LDCL, and delta2-agonist deltorphin 2 promoted 15 mkg/ml zymosan-induced LDCL only toward the 25 minutes of the experiment. beta-endorphin and selective d- and m- agonists did not affect the spontaneous and suppressed 15 mkg/ml and 150 mkg/ml zymosan-induced neutrophil LDCL. Therefore, opioid peptides play essential role in the process of direct and indirect regulation of oxygen-dependent system of neutrophil granulocyte bactericidal activity.
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
Sigma-Aldrich
[D-Ala2]-Deltorphin II, ≥97% (HPLC)