Merck
  • Unrestricted Hartree-Fock based on the fragment molecular orbital method: energy and its analytic gradient.
Unrestricted Hartree-Fock based on the fragment molecular orbital method: energy and its analytic gradient.
The Journal of chemical physics (2012-08-03)
Hiroya Nakata, Dmitri G Fedorov, Takeshi Nagata, Satoshi Yokojima, Koji Ogata, Kazuo Kitaura, Shinichiro Nakamura
ABSTRACT
A consideration of the surrounding environment is necessary for a meaningful analysis of the reaction activity in large molecular systems. We propose an approach to perform unrestricted Hartree-Fock (UHF) calculations within the framework of the fragment molecular orbital (FMO) method (FMO-UHF) to study large systems with unpaired electrons. Prior to an energy analysis one has to optimize geometry, which requires an accurate analytic energy gradient. We derive the FMO-UHF energy and its analytic gradient and implement them into GAMESS. The performance of FMO-UHF is evaluated for a solvated organic molecule and a solvated metal complex, as well as for the active part of a protein, in terms of energy, gradient, and geometry optimization.
Sigma-Aldrich
TEMPO, 98%
Sigma-Aldrich
TEMPO, purified by sublimation, 99%
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%
Sigma-Aldrich
Manganese(II) acetylacetonate
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) acetylacetonate, 97%