Merck
E versus Z diazeniumdiolation of acetoacetate-derived carbanions.
The Journal of organic chemistry (2012-08-03)
Navamoney Arulsamy, D Scott Bohle, Carla L Holman, Inna Perepichka
ABSTRACT
Nitric oxide adds to methyl acetoacetate in the presence of KOH in methanol at room temperature to form potassium acetylsydnonate N-oxide (K1) with an (E)-diazeniumdiolation and potassium acetate diazenium diolate (K(2)2) from a (Z)-diazeniumdiolation. A study of the reaction with LiOH, NaOH, and NMe(4)OH and with ethyl acetate substrate reveals that the temperature of the reaction greatly influences the nitric oxide reactivity. At 23 °C, nitric oxide adds to give both E and Z products, whereas at -5 °C the gas reacts almost exclusively to give Z addition. The (Z)-diazeniumdiolation products, namely, the alkali metal and NMe(4)(+) salts of methyl and ethylbutenoate-2-diazeniumdiolate-3-hydroxylate (3(2-) and 4(2-)), are isolated in good yields. The alkali metal salts are not amenable for recrystallization because of their ready decomposition in aqueous solutions. However, [NMe(4)](2)[MeC(O)C(N(2)O(2))CO(2)Me] is readily recrystallized from a methanol/acetonitrile solvent mixture. The crystals are unambiguously characterized by X-ray crystallography. NMR spectra for all of the 3(2-) and 4(2-) salts reveal the presence of two isomers in aq solutions. But the structure of the NMe(4)(+) salt contains only one of the isomers. Our attempts to cyclize the isolated and purified butenoatediazeniumdiolates from the (Z)-diazeniumdiolation to the E-containing sydnonate products were unsuccessful. TGA/DSC data for all of the products demonstrate the thermal instability of the salts at high temperatures. The salts decompose exothermally possibly with the release of N(2)O among other gases.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, ≥98.5% (GC)
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O