Merck
New cost-effective method for conversion of vegetable oil to biodiesel.
Bioresource technology (2012-08-04)
K D Pal, A Prakash
ABSTRACT
A low cost method to convert triglycerides to biodiesel by transesterification reaction has been investigated. The new approach is based on controlled feeding of oil into a pool of alcohol allowing easy dispersion of oil creating large interfacial area for mass transfer. It also enabled equilibrium constants to shift in the forward direction due to excess of alcohol in the dispersion. As a result product yield higher than 95% could be achieved at lower energy input compared to conventional method. Comparisons made with conventional batch and co-solvent enabled method show differences and highlight improvements. Reactions were conducted in one liter reactor at various residence time, temperature and molar ratio of canola oil with methanol in presence of potassium hydroxide as the catalyst. Samples were collected at regular intervals and analyzed by gas chromatography (GC) to determine the reaction progress.
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC