Merck
  • Synthesis of eugenol esters by lipase-catalyzed reaction in solvent-free system.
Synthesis of eugenol esters by lipase-catalyzed reaction in solvent-free system.
Applied biochemistry and biotechnology (2012-08-07)
V Chiaradia, N Paroul, R L Cansian, C V Júnior, M R Detofol, L A Lerin, J Vladimir Oliveira, D Oliveira
ABSTRACT
Enzymatic esterification of eugenol is a matter of great scientific and technological interest due to the well-known drawbacks of the chemical-catalyzed route as well as the potential use of produced compounds as natural antimicrobials. This work reports the maximization of eugenil acetate production by esterification of eugenol and acetic anhydride in a solvent-free system using Novozym 435 as catalyst. The antimicrobial activity of eugenol and eugenil acetate was also determined. The operating conditions that maximized eugenil acetate production were 50 °C, eugenol to acetic anhydride of 1:3, 150 rpm, and 5.5 wt% of enzyme, with a conversion of 99 %. A kinetic study was performed to assess the influence of substrates molar ratio, enzyme concentration, and temperature on eugenil acetate yield. Results show that an excess of anhydride, low enzyme concentration (1 wt%), and 60 °C afforded nearly complete conversion after 6 h of reaction. The highest antimicrobial activity of eugenil acetate was observed against Acinetobacter sp. (48.66 mm) at concentration of 20 μL. Results indicate that the esterification of eugenol improved its antimicrobial properties. New experimental data on enzymatic esterification of eugenol and acetic anhydride are reported in this work, showing a promising perspective to overcome the inconvenient of the chemical-catalyzed route for obtaining antimicrobial natural compounds.
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Supelco
Acetic anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)