Merck
  • The effects of sodium diethyldithiocarbamate in fibroblasts V79 cells in relation to cytotoxicity, antioxidative enzymes, glutathione, and apoptosis.
The effects of sodium diethyldithiocarbamate in fibroblasts V79 cells in relation to cytotoxicity, antioxidative enzymes, glutathione, and apoptosis.
Archives of toxicology (2012-08-09)
I Rahden-Staroń, E Grosicka-Maciąg, D Kurpios-Piec, H Czeczot, T Grzela, M Szumiło
ABSTRACT
Sodium diethyldithiocarbamate (DETC) is the main metabolite of disulfiram. Recently, we reported that mechanism of disulfiram cytotoxicity in V79 cells might be partially connected with thiol redox-state imbalance. Here, we examined the effect of DETC on the level of intracellular glutathione (GSH), protein oxidation (measured as PC-protein carbonyl content), lipid peroxidation (measured as TBARS-thiobarbituric acid reactive substances), antioxidant enzymatic defense, as well as on apoptosis. We used V79 Chinese hamster fibroblasts cells with and without modulated glutathione (GSH) level by N-acetyl-L-cysteine (NAC). We showed that treatment with DETC at concentrations that cause a moderate increase in thiol-state imbalance but not cell death stimulates oxidative stress measured as increased level of PC and TBARS, adaptive response of GSH-related enzymes and apoptosis. Our results show that cellular effects of DETC are partially attributable to the initial redox cellular state, since the increase of GSH level by NAC pre-treatment prevented the observed changes.
Product Description
Disulfiram, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate, ACS reagent
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate
Sigma-Aldrich
Ammonium diethyldithiocarbamate
Supelco
Sodium diethyldithiocarbamate trihydrate, ACS reagent, for the determination and separation of heavy metals, ≥97%
Sigma-Aldrich
Tetraethylthiuram disulfide, ≥97.0% (S)