Merck
  • Synthesis and anti-inflammatory effects of new piperazine and ethanolamine derivatives of H(1)-antihistaminic drugs.
Synthesis and anti-inflammatory effects of new piperazine and ethanolamine derivatives of H(1)-antihistaminic drugs.
Mini reviews in medicinal chemistry (2012-08-11)
Abbas Ahmadi, Mohsen Khalili, Ali Nafarie, Arash Yazdani, Babak Nahri-Niknafs
ABSTRACT
In addition to their antihistamine effects, H1-receptor antagonists possess pharmacological properties that are not uniformly distributed among this class of drugs, such as anti-inflammatory, anti-allergic and antiplatelet activities. In this paper, Cyclizine (1-benzhydryl-4-methyl-piperazine, I), bromodiphenhydramine (2-[(4-bromophenyl)-phenylmethoxy]-N, N-dimethylethanamine, II) and some of their new piperazine and ethanolamine derivatives (III-VIII) inducing changes in substitution of phenyl and amine moieties were synthesized and their acute and chronic antiinflammatory effects were evaluated by standard pharmacological tests. The results showed that substitution of phenyl by tolyl, anisol and cumene groups in piperazine family could remarkably decrease acute inflammation in these new drugs. Also, substitution of dimethylamine by morpholine group could not decrease this inflammation in new synthesized ethanolamine family. But the results from the cotton pellet-induced granuloma formation in rats showed that none of drugs (I-VIII) were effective to reduce the chronic inflammation.
Product Description
Supelco
Diphenhydramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Ethanolamine, ≥98%
Supelco
Piperazine hexahydrate, analytical standard
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Diphenhydramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
Supelco
Diphenhydramine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%