Merck
Single-stranded siRNAs activate RNAi in animals.
Cell (2012-09-04)
Walt F Lima, Thazha P Prakash, Heather M Murray, Garth A Kinberger, Wenyu Li, Alfred E Chappell, Cheryl S Li, Susan F Murray, Hans Gaus, Punit P Seth, Eric E Swayze, Stanley T Crooke
ABSTRACT
The therapeutic utility of siRNAs is limited by the requirement for complex formulations to deliver them to tissues. If potent single-stranded RNAs could be identified, they would provide a simpler path to pharmacological agents. Here, we describe single-stranded siRNAs (ss-siRNAs) that silence gene expression in animals absent lipid formulation. Effective ss-siRNAs were identified by iterative design by determining structure-activity relationships correlating chemically modified single strands and Argonaute 2 (AGO2) activities, potency in cells, nuclease stability, and pharmacokinetics. We find that the passenger strand is not necessary for potent gene silencing. The guide-strand activity requires AGO2, demonstrating action through the RNAi pathway. ss-siRNA action requires a 5' phosphate to achieve activity in vivo, and we developed a metabolically stable 5'-(E)-vinylphosphonate (5'-VP) with conformation and sterioelectronic properties similar to the natural phosphate. Identification of potent ss-siRNAs offers an additional option for RNAi therapeutics and an alternate perspective on RNAi mechanism.
Sigma-Aldrich
Vinylphosphonic acid, 97%
Sigma-Aldrich
Vinylphosphonic acid, ≥90% (T)