Merck
Enantioselective inhibition of dichlorprop on catalase.
Bulletin of environmental contamination and toxicology (2012-09-11)
Yun Ma, Jihong Jiang, Chao Xu, Xianting Lu
ABSTRACT
The enantioselectivity interaction of 2,4-dichlorprop (DCPP) and catalase were studied, and it was further evaluated with the presence of humus. Both of rac-DCPP and R-DCPP can inhibit the activity of catalase with the concentrations of 0.05-80 mg L(-1), the inhibitory type of rac-DCPP was uncompetitive, and of R-DCPP was complex. The presence of humic acid has changed the inhibitory ability of DCPP on catalase, the inhibition of rac-DCPP disappeared and the inhibition type of R-DCPP mainly became uncompetitive. These results suggest that inhibition of chiral DCPP on catalase is enantioselective.
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, 97%
Supelco
Dichlorprop, PESTANAL®, analytical standard
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
2,4-D, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland