Merck
  • Highly enantioselective and recyclable organocatalytic Michael addition of malonates to α,β-unsaturated aldehydes in aqueous media.
Highly enantioselective and recyclable organocatalytic Michael addition of malonates to α,β-unsaturated aldehydes in aqueous media.
Organic & biomolecular chemistry (2012-09-15)
Subrata K Ghosh, Kritanjali Dhungana, Allan D Headley, Bukuo Ni
ABSTRACT
A new type of pyrrolidine-based organocatalyst, which was developed earlier in our lab, has been found to be very effective for the Michael addition reaction in aqueous solvents involving a wide range of α,β-unsaturated aldehydes and malonate derivatives. For the reactions studied, good to excellent yields (73%-96%) and high to excellent enantioselectivities (up to 97%) were obtained using this catalyst. In addition, the catalyst could be recycled up to four times with gradual reductions in yields and enantioselectivity observed after the second cycle.
Sigma-Aldrich
Sodium malonate dibasic, ≥97.0% (NT)
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Pyrrolidine, 99%
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Supelco
Malonic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland