Merck
  • Direct benzylic metalation of a phenethylamine derivative: potassium as the key to both generation and stabilization of a labile anion.
Direct benzylic metalation of a phenethylamine derivative: potassium as the key to both generation and stabilization of a labile anion.
Chemical communications (Cambridge, England) (2012-09-21)
Christian Unkelbach, Hannah S Rosenbaum, Carsten Strohmann
ABSTRACT
t-BuLi-t-BuOK selectively metalates the benzylic position of 2-phenylethyldimethylamine under mild conditions without occurrence of β-elimination in the resulting metalated species. Theoretical and structural studies indicate that potassium is crucial for both the lowering of the barrier of the initial deprotonation step and the stabilization of the labile anion.
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane
Sigma-Aldrich
n-Butyllithium solution, 1.6 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.7 M in heptane
Sigma-Aldrich
2-Phenylethylamine hydrochloride, ≥98%
Sigma-Aldrich
n-Butyllithium solution, 11.0 M in hexanes
Sigma-Aldrich
Phenethylamine, 99%
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Phenethylamine, ≥99%
Supelco
2-Phenethylamine, analytical standard