Merck
  • Vaginal atrophy in breast cancer survivors: role of vaginal estrogen therapy.
Vaginal atrophy in breast cancer survivors: role of vaginal estrogen therapy.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology (2012-09-22)
Luciano Mariani, Angiolo Gadducci, Enrico Vizza, Silverio Tomao, Patrizia Vici
ABSTRACT
Early menopause and related vaginal atrophy is a well known side-effect of hormone adjuvant treatment in breast cancer patients, particularly during aromatase-inhibitors therapy. Due to estrogens contra-indication, proper therapy for such symptom remains often an inadequately addressed clinical problem. After an accurate assessment of the risk/benefit ratio, vaginal low-dose estrogen treatment (better with estriol) [corrected] may have a role in controlling vaginal atrophy in selected and informed breast cancer women.
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Estriol, meets USP testing specifications
Supelco
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®