Merck
  • Simultaneous determination of zopiclone and its degradation product and main impurity (2-amino-5-chloropyridine) by micellar liquid chromatography with time-programmed fluorescence detection: preliminary investigation for biological monitoring.
Simultaneous determination of zopiclone and its degradation product and main impurity (2-amino-5-chloropyridine) by micellar liquid chromatography with time-programmed fluorescence detection: preliminary investigation for biological monitoring.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-09-25)
R N El-Shaheny, A Alattas, J J Nasr, N El-Enany, F Belal
ABSTRACT
A simple and reliable HPLC method was developed and validated for the simultaneous determination of the hypnotic drug, zopiclone (ZPC) and its degradation product and main impurity, 2-amino-5-chloropyridine (ACP). The analyses were carried out on BDS Hypersil phenyl column (4.6 mm × 250 mm, 5 μm particle size) using micellar mobile phase consisting of 0.15M SDS, 10% n-propanol, 0.3% triethylamine, and 0.02 M orthophosphoric acid (pH 3.5) with timed programmable fluorescence detection. The proposed method was found to be rectilinear over the concentration ranges of 0.5-10.0 μg/mL for ZPC and 2.5-50 ng/mL for ACP. Moreover, the method was applied for the determination of ZPC in commercial tablets with mean percentage recovery of 99.06±1.49. The results of the proposed method were statistically compared with those obtained by the comparison method revealing no significance differences in the performance of the two methods regarding accuracy and precision. Furthermore, the proposed method was applied for the detection and determination of ACP in human urine as a marker for ZPC intake.
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
2-Amino-5-chloropyridine, 98%
Supelco
Zopiclone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®