Merck
  • Toxicology and carcinogenesis studies of N,N-dimethyl-p-toluidine (CAS No. 99-97-8) in F344/N rats and B6C3F1/N mice (gavage studies).
Toxicology and carcinogenesis studies of N,N-dimethyl-p-toluidine (CAS No. 99-97-8) in F344/N rats and B6C3F1/N mice (gavage studies).
National Toxicology Program technical report series (2012-10-03)
ABSTRACT
N,N-dimethyl-p-toluidine was nominated for toxicology and carcinogenesis studies by the National Cancer Institute based on the potential for human exposure through its use in dental materials and bone cements and the lack of toxicity and carcinogenicity data. Male and female F344/N rats and B6C3F1/N mice were administered N,N-dimethyl-p-toluidine (greater than 99% pure) in corn oil by gavage for 3 months or 2 years. Genetic toxicology studies were conducted in Salmonella typhimurium and Escherichia coli, mouse peripheral blood, and mouse and rat liver. 3-MONTH STUDY IN RATS: Groups of 10 male and 10 female rats were administered 0, 62.5, 125, 250, 500, or 1,000 mg N,N-dimethyl-p-toluidine/kg body weight in corn oil by gavage, 5 days per week for 14 weeks. Additional groups of 10 male and 10 female rats (clinical pathology study) were administered the same doses, 5 days per week for 25 days. On day 88, blood was collected from core study rats for hemoglobin and methemoglobin analyses only. All 1,000 mg/kg male and female rats and one 500 mg/kg male rat died by study day 3. Mean body weights of all surviving dosed groups of males and females were significantly less than those of the vehicle controls. Clinical findings associated with exposure to N,N-dimethyl-p-toluidine included cyanosis, abnormal breathing, and lethargy in groups administered 250 mg/kg or greater. Methemoglobinemia appeared to be the primary hematologic toxic response, and many other lesions could be explained as secondary to methemoglobin formation including Heinz body formation; a macrocytic, hypochromic, responsive anemia; and increased hematopoietic cell proliferation in the spleen and bone marrow. In general, hematologic changes were dose-related and occurred at both evaluated timepoints in all dosed groups. Anemia was evidenced by decreases in hematocrit values, hemoglobin concentrations, and erythrocyte counts; erythrocyte macrocytosis was characterized by increases in mean cell volume and mean cell hemoglobin values; erythrocyte hypochromia was evidenced by decreases in mean cell hemoglobin concentration values; and an erythropoietic response to the anemia was characterized by substantially increased reticulocyte and nucleated erythrocyte counts. Liver weights of all surviving dosed groups of males and females were significantly greater than those of the vehicle controls. Kidney weights of all surviving dosed groups of females were significantly greater than those of the vehicle controls. There were significant decreases in left cauda epididymis and left epididymis weights in 250 mg/kg males. There was a dose-related decrease in the number of cycling females, with only four females in the 250 mg/kg group having regular cycles and females in the 125 and 250 mg/kg groups spending a significantly higher proportion of time in extended diestrus compared to the vehicle control group. In the surviving groups of rats, there were significantly increased incidences of pigmentation in the liver of all dosed groups, hepatocyte hypertrophy in groups administered 125 mg/kg or greater, and hepatocyte necrosis in 62.5, 250, and 500 mg/kg females. In the olfactory epithelium of the nose, there were dose-related increases in the incidences and severities of degeneration in all dosed groups and significantly increased incidences of metaplasia in the 250 and 500 mg/kg groups. In the respiratory epithelium of the nose, there were significantly increased incidences of hyperplasia and squamous metaplasia in all of the groups administered 125 mg/kg or greater. The incidences of glandular hyperplasia of the nose were significantly increased in males and females administered 125, 250, or 500 mg/kg. In the spleen, there were significantly increased incidences of capsule fibrosis, congestion, mesothelial hypertrophy, and lymphoid follicle atrophy primarily in groups administered 125 mg/kg or greater. Hematopoietic cell proliferation and pigmentation were increased in severity in treated groups. In the kidney, there were significantly increased incidences of nephropathy (females), pigmentation (males and females), papillary necrosis (males and females), and mineralization (males). Other treatment-related lesions included inflammation of the forestomach in males, mesenteric lymph node atrophy in females, and bone marrow hyperplasia in males and females. 3-MONTH STUDY IN MICE: Groups of 10 male and 10 female mice were administered 0, 15, 30, 60, 125, or 250 mg N,N-dimethyl-p-toluidine/kg body weight in corn oil by gavage, 5 days per week for 14 weeks. All 250 mg/kg male and female mice (except for one male mouse) died before day 10, and three males and two females administered 125 mg/kg died before the end of the study. The final mean body weight of 125 mg/kg males and the mean body weight gains of 125 mg/kg males and females were significantly less than those of the vehicle controls. Clinical findings associated with administration of N,N-dimethyl-p-toluidine included abnormal breathing, thinness, lethargy, cyanosis, and ruffled fur in 125 and 250 mg/kg males and females. Methemoglobinemia appeared to be the primary hematologic toxic response; however there were less severe erythron changes compared to the 3-month study in rats. In females, no erythron changes were detected up to 125 mg/kg. In males, inconsistent and minor decreases in hematocrit values, hemoglobin concentrations, and erythrocyte counts, and increased reticulocyte counts occurred in groups administered 60 mg/kg or greater. Methemoglobin values were minimally increased in males and females administered 30 mg/kg or greater. Heinz bodies were slightly increased in 60 mg/kg females, 125 mg/kg males and females, and the one surviving 250 mg/kg male; Heinz body formation was considered secondary to methemoglobin formation. Liver weights of all dosed groups of mice were significantly greater than those of the vehicle controls. In the surviving groups of mice, there were significantly increased incidences of bronchiolar epithelium degeneration, bronchiolar epithelium regeneration, and peribronchiolar chronic active inflammation in the lung of 125 mg/kg groups, and histiocytic infiltrates of the alveoli in 125 mg/kg females. In the nose, there were significantly increased incidences of glandular hyperplasia and olfactory epithelium metaplasia in the 125 mg/kg groups and olfactory epithelium degeneration in 60 mg/kg females and 125 mg/kg males and females. In the thymus, the incidences of thymocyte necrosis in the 125 mg/kg groups were significantly increased. In the liver, the severities of cytoplasmic vacuolization of the hepatocytes were increased in dosed groups of males and females. 2-YEAR STUDY IN RATS: Groups of 50 male and 50 female rats were administered 0, 6, 20, or 60 mg N,N-dimethyl-p-toluidine/kg body weight in corn oil by gavage, 5 days per week for 104 or 105 weeks. Additional groups of 10 male and 10 female rats (clinical pathology study) were administered the same doses for 86 days. Survival of 60 mg/kg males was significantly less than that of the vehicle controls. Mean body weights of 60 mg/kg males and females were more than 10% less than those of the vehicle controls after week 61 and week 33, respectively. Clinical findings included signs of pallor in 60 mg/kg females and hyperactivity and boxing behavior in 20 mg/kg females and 60 mg/kg males and females. The hematology findings at the 3-month timepoint were consistent with those in the 3-month study in rats which indicated that methemoglobinemia was the primary hematologic toxic response. In the 20 and 60 mg/kg groups, there were dose-related decreases in hematocrit values, hemoglobin concentrations, and erythrocyte counts. There were similar trends toward erythrocyte macrocytosis and hypochromia and increased erythropoiesis as seen in the 3-month study. While the magnitudes of the erythron decreases were not sufficient to classify the responses as anemias, the patterns of the erythron changes were identical to those in the 3-month study. In the liver of 60 mg/kg males and females, there were significantly increased incidences of hepatocellular carcinoma and hepatocellular adenoma or hepatocellular carcinoma (combined). Numerous nonneoplastic liver lesions occurred in dosed males and females primarily in the 20 and 60 mg/kg groups. In the nose, there were significantly increased incidences of transitional epithelium adenoma and transitional epithelium adenoma or carcinoma (combined) in 60 mg/kg males; transitional epithelium adenoma also occurred in female rats administered 6 or 60 mg/kg. In the nose, there were significantly increased incidences of nonneoplastic lesions in the olfactory, respiratory, and transitional epithelia of dosed rats. These lesions occurred with the greatest incidence and severity in the 60 mg/kg groups. The incidences of inflammation and nerve atrophy were significantly increased in males and females administered 60 mg/kg. There were increased incidences of follicular cell adenoma or carcinoma (combined) of the thyroid gland in all dosed groups of males, and an increased incidence of follicular cell adenoma in 20 mg/kg females. In the spleen, there were significantly increased incidences of hematopoietic cell proliferation in all dosed groups of males and females. The incidences of congestion and mesothelial hypertrophy of the capsule were significantly increased in 60 mg/kg males and all dosed groups of females. There were also significantly increased incidences of capsular fibrosis and atrophy of the lymphoid follicle in the 60 mg/kg groups. The incidences of pigmentation were significantly increased in all dosed groups of males and in 60 mg/kg females. In all dosed groups of female rats, there were significantly increased incidences of nephropathy. (ABSTRACT TRUNCATED)
Sigma-Aldrich
4,N,N-Trimethylaniline, 99%
Sigma-Aldrich
4,N,N-Trimethylaniline, ≥98.5% (GC)
Sigma-Aldrich
4,N,N-Trimethylaniline, purum, ≥98.0% (GC)