Merck
  • Is dual morphology of rock-salt crystals possible with a single additive? The answer is yes, with barbituric acid.
Is dual morphology of rock-salt crystals possible with a single additive? The answer is yes, with barbituric acid.
Angewandte Chemie (International ed. in English) (2012-10-06)
Anik Sen, Bishwajit Ganguly
ABSTRACT
Crystal face lift: barbituric acid is shown to be a new crystal-habit modifier for sodium chloride crystals. Two morphologies of salt crystals can be prepared separately with this new additive. It is of the few additives able to induce rhombic dodecahedron crystals for NaCl, and is required only a trace of amount, unlike other additives, such as glycine.
Sigma-Aldrich
Sodium barbiturate, ≥97.0% (T)
Supelco
Barbituric acid, for spectrophotometric det. of cyanide, ≥99.5%
Sigma-Aldrich
Barbituric acid, ReagentPlus®, 99%