Merck
  • Turning the volume down on heavy metals using tuned diatomite. A review of diatomite and modified diatomite for the extraction of heavy metals from water.
Turning the volume down on heavy metals using tuned diatomite. A review of diatomite and modified diatomite for the extraction of heavy metals from water.
Journal of hazardous materials (2012-10-16)
Angela F Danil de Namor, Abdelaziz El Gamouz, Sofia Frangie, Vanina Martinez, Liliana Valiente, Oliver A Webb
ABSTRACT
Contamination of water by heavy metals is a global problem, to which an inexpensive and simple solution is required. Within this context the unique properties of diatomite and its abundance in many regions of the world have led to the current widespread interest in this material for water purification purposes. Defined sections on articles published on the use of raw and modified diatomite for the removal of heavy metal pollutants from water are critically reviewed. The capability of the materials as extracting agents for individual species and mixtures of heavy metals are considered in terms of the kinetics, the thermodynamics and the recyclability for both, the pollutant and the extracting material. The concept of 'selectivity' for the enrichment of naturally occurring materials such as diatomite through the introduction of suitable functionalities in their structure to target a given pollutant is emphasised. Suggestions for further research in this area are given.
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Diatomaceous earth
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Supelco
Celite® 545 AW, reagent grade
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate