Merck
  • Suppression of pain-related behavior in two distinct rodent models of peripheral neuropathy by a homopolyarginine-conjugated CRMP2 peptide.
Suppression of pain-related behavior in two distinct rodent models of peripheral neuropathy by a homopolyarginine-conjugated CRMP2 peptide.
Journal of neurochemistry (2012-10-31)
Weina Ju, Qi Li, Yohance M Allette, Matthew S Ripsch, Fletcher A White, Rajesh Khanna
ABSTRACT
The N-type voltage-gated calcium channel (CaV2.2) is a clinically endorsed target in chronic pain treatments. As directly targeting the channel can lead to multiple adverse side effects, targeting modulators of CaV2.2 may prove better. We previously identified ST1-104, a short peptide from the collapsin response mediator protein 2 (CRMP2), which disrupted the CaV2.2-CRMP2 interaction and suppressed a model of HIV-related neuropathy induced by anti-retroviral therapy but not traumatic neuropathy. Here, we report ST2-104 -a peptide wherein the cell-penetrating TAT motif has been supplanted with a homopolyarginine motif, which dose-dependently inhibits the CaV2.2-CRMP2 interaction and inhibits depolarization-evoked Ca(2+) influx in sensory neurons. Ca(2+) influx via activation of vanilloid receptors is not affected by either peptide. Systemic administration of ST2-104 does not affect thermal or tactile nociceptive behavioral changes. Importantly, ST2-104 transiently reduces persistent mechanical hypersensitivity induced by systemic administration of the anti-retroviral drug 2',3'-dideoxycytidine (ddC) and following tibial nerve injury (TNI). Possible mechanistic explanations for the broader efficacy of ST2-104 are discussed.
Sigma-Aldrich
Poly-L-arginine hydrochloride, mol wt >70,000
Sigma-Aldrich
Poly-L-arginine hydrochloride, mol wt 15,000-70,000
Sigma-Aldrich
Poly-L-arginine hydrochloride, mol wt 5,000-15,000