Merck
  • Vascular malformation of the glans penis successfully treated with Nd:YAG laser.
Vascular malformation of the glans penis successfully treated with Nd:YAG laser.
Dermatology online journal (2012-11-06)
Verónica López, José M Ricart, Inmaculada López, José M Martín, Davide Marton, Carmen Ortega, Salvador Costa
ABSTRACT
Penile venous malformations are uncommon and those located on the glans are even rarer. Treatment of venous malformations of the glans is currently controversial. Neodynium (Nd):YAG laser treatment may be a useful option.
Product Description
Oxytetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oxytetracycline dihydrate, Assay (anhydrous basis) 95.0%-102.0%
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Polymyxin B sulfate salt
Supelco
Oxytetracycline hydrochloride, VETRANAL®, analytical standard
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Sigma-Aldrich
Oxytetracycline hydrochloride
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications
Millipore
Oxytetra Selective Supplement, suitable for microbiology
Polymyxin B sulfate, European Pharmacopoeia (EP) Reference Standard