Merck
Bioactive constituents of Salvia chrysophylla Stapf.
Natural product research (2012-11-07)
Burcu Çulhaoğlu, Gönül Yapar, Tuncay Dirmenci, Gülaçtı Topçu
ABSTRACT
The dichloromethane extract of the aerial parts of Salvia chrysophylla Stapf (Lamiaceae), which is an endemic species to south-western Anatolia, was studied for non-volatile secondary metabolites for the first time in this study. Structures of the isolated compounds were elucidated as sclareol, β-sitosterol, salvigenin, oleanolic acid and ursolic acid. The lipid peroxidation inhibitory activity and the DPPH free radical scavenging activity of the pure isolates were investigated to establish their antioxidant potential. Their anticholinesterase activity was carried out by the Ellman assay against both enzymes, acetylcholinesterase (AChE) and butyrylcholinesterase, and diterpene sclareol exhibited fairly good activity against both the enzymes while the two triterpenoids oleanolic and ursolic acids exhibited selective activity against AChE.
Sigma-Aldrich
Sclareol, 98%
Supelco
Ursolic acid, analytical standard
Sigma-Aldrich
Ursolic acid, ≥90%
Ursolic acid, primary reference standard
Supelco
Dichloromethane, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer