Merck
  • Analytical performance of a lab-made concomitant metal analyzer to generate volatile species of Ag, Au, Cd, Cu, Ni, Sn and Zn using 8-hydroxyquinoline as a reaction media.
Analytical performance of a lab-made concomitant metal analyzer to generate volatile species of Ag, Au, Cd, Cu, Ni, Sn and Zn using 8-hydroxyquinoline as a reaction media.
Talanta (2012-11-13)
Julia Villanueva-Alonso, Elena Peña-Vázquez, Pilar Bermejo-Barrera
ABSTRACT
This study evaluated the main parameters affecting Ag, Au, Cd, Cu, Ni, Sn and Zn vapor generation using a lab-made concomitant metal analyzer (CMA) as a reaction chamber and gas-liquid separator. The modifier used in the reaction media was 8-hydroxyquinoline, and Inductively-Coupled Plasma Optical Emission Spectrometry was used as detection technique. The performance of the lab-made concomitant analyzer was compared with the performance of a continuous flow gas-liquid separator and of a cyclonic spray chamber. Standards were prepared in acid media and included 1 mg L(-1) of Co as a catalyzer. The optimum concentrations of the reagents in the standards were: 450 mg L(-1) of 8-hydroxyquinoline and 0.4 M nitric acid. The optimum concentration of sodium borohydride to generate the vapors was 2.25% (w/v) (prepared in 0.4% (w/v) NaOH). The volatile species were swept from the CMA to the torch by an argon flow of 0.6 mL min(-1). The use of the CMA led to an improvement of the detection limits for some elements compared to conventional nebulization: 1.1 μg L(-1) for Ag, 7.0 μg L(-1) for Au and 4.3 μg L(-1) for Sn. The limit of detection for Cu was 1.4 μg L(-1) and for Ni 22.5 μg L(-1). The direct mixing of the reagents on the spray chamber was not effective for Cd and Zn; a deviation of the linearity was observed for these elements.
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Supelco
8-Quinolinol, PESTANAL®, analytical standard
Sigma-Aldrich
8-Quinolinol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (perchloric acid titration)
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
8-Hydroxyquinoline
Sigma-Aldrich
8-Hydroxyquinoline, ACS reagent, 98.5%