Merck
  • Ionic matrices pre-spotted matrix-assisted laser desorption/ionization plates for patient maker following in course of treatment, drug titration, and MALDI mass spectrometry imaging.
Ionic matrices pre-spotted matrix-assisted laser desorption/ionization plates for patient maker following in course of treatment, drug titration, and MALDI mass spectrometry imaging.
Analytical biochemistry (2012-11-13)
David Bonnel, Julien Franck, Céline Mériaux, Michel Salzet, Isabelle Fournier
ABSTRACT
In the current study, we compared plastic matrix-assisted laser desorption/ionization (MALDI) plates pre-spotted with different solid ionic matrices. Data reflect that after 3 months of storage, the standards were oxidized in α-cyano-4-hydroxycinnamic acid (HCCA) whether or not in HCCA/3-acetylpyridine (3APY) and HCCA/aniline, and certain peptides, such as ubiquitin, were not detected using the HCCA matrix, whereas they were detected in pre-spotted ionic matrices. Application in peptidomics of these MALDI matrices pre-spotted plates (after 3 months of storage) with ovarian cyst fluid showed less intense signals with HCCA than with solid ionic matrices. We show that these pre-spotted ionic matrices plates can be used for relative drug quantification, high mass protein detection, and MALDI mass spectrometry imaging.
Sigma-Aldrich
3-Acetylpyridine, 98%
Sigma-Aldrich
3-Acetylpyridine, ≥98%, FG
Sigma-Aldrich
Aniline hydrochloride, ≥99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, Ultra pure
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Supelco
Atenolol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®