Merck
  • Tube radial distribution of solvents observed in an aqueous ionic liquid mixed solution delivered into a capillary tube.
Tube radial distribution of solvents observed in an aqueous ionic liquid mixed solution delivered into a capillary tube.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2012-11-15)
Yuki Kawai, Masaharu Murata, Masahiko Hashimoto, Kazuhiko Tsukagoshi
ABSTRACT
The tube radial distribution of solvents was observed in an aqueous ionic liquid mixed solution of 1-butyl-3-methylimidazolium chloride fed into a capillary tube. A phase diagram was constructed with 1-butyl-3-methylimidazolium chloride and potassium hydroxide, which included boundary curves between homogeneous and heterogeneous solutions at 15 and 20°C. As an example, an aqueous ionic liquid mixed homogeneous solution, comprising the ratio of 1-butyl-3-methylimidazolium chloride:potassium hydroxide (30.5:16.5 wt/wt%), which were positioned near the boundary at 20°C, were delivered into a fused-silica capillary tube (75 μm i.d., 110 cm length, 15°C tube temperature) at a flow rate of 1.0 μL min(-1). The homogeneous solution changed to a heterogeneous solution with two phases: the inner (the aqueous phase merely containing the ionic liquid) and the outer (the ionic liquid-rich) phases, in the capillary tube. The radial distribution of the solvents in the aqueous ionic liquid mixed solution was observed through a bright-light microscope-CCD camera system.
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC