Merck
Pharmacokinetic interaction between methotrexate and chloral hydrate.
Pediatric blood & cancer (2012-11-16)
Kim Dao, Anton Ivanyuk, Thierry Buclin, Maja Beck-Popovic, Manuel Diezi
ABSTRACT
We report the case of a drug interaction between methotrexate (MTX) and chloral hydrate (CH) observed in a child treated for acute leukemia. Significantly slower MTX clearance and increased MTX exposure occurred on the first three courses of a high-dose chemotherapy when co-administered with CH despite normal renal function, adequate hydration, and alkalinization. Mean MTX area under the curve associated with CH administration was 1,134 µmol hours/L, compared to 608 µmol hours/L after discontinuation of CH. This interaction possibly resulted from a competition between anionic CH metabolites and MTX for renal tubular excretion.
Sigma-Aldrich
Chloral hydrate, ≥99%
Sigma-Aldrich
Chloral hydrate, crystallized, ≥98.0% (T)