Merck
  • A distal renal tubular acidosis showing hyperammonemia and hyperlactacidemia.
A distal renal tubular acidosis showing hyperammonemia and hyperlactacidemia.
La Pediatria medica e chirurgica : Medical and surgical pediatrics (2012-11-24)
C Ripoli, A Pinna, S Marras, M L Fenu, A M Nurchi
ABSTRACT
distal renal tubular acidosis (dRTA) presents itself with variable clinical manifestations and often with late expressions that impact on prognosis. A 45-day-old male infant was admitted with stopping growth, difficult feeding and vomiting after meals. Clinical tests and labs revealed a type 1 renal tubular acidosis, even if the first blood tests showed ammonium and lactate increase. We had to exclude metabolic diseases before having a certain diagnosis. blood and urine investigations and genetic tests are fundamental to formulate dRTA diagnosis and to plan follow-up, according to possible phenotypic expressions of recessive and dominant autosomal forms in patients with dRTA.
Sigma-Aldrich
Potassium citrate tribasic monohydrate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets USP testing specifications
Sigma-Aldrich
Potassium citrate tribasic monohydrate, suitable for cell culture
Sigma-Aldrich
Potassium citrate tribasic monohydrate, ≥98% (GC/titration)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets analytical specification of Ph. Eur., BP, FCC, E332, 99-100.5% (calc with ref. to anhyd. subst.)