Merck
  • ABC triblock terpolymers exhibiting both temperature- and pH-sensitive micellar aggregation and gelation in aqueous solution.
ABC triblock terpolymers exhibiting both temperature- and pH-sensitive micellar aggregation and gelation in aqueous solution.
Langmuir : the ACS journal of surfaces and colloids (2012-11-30)
Isha Koonar, Can Zhou, Marc A Hillmyer, Timothy P Lodge, Ronald A Siegel
ABSTRACT
Two poly(ethylene-alt-propylene)-b-poly(ethylene oxide)-b-poly(N-isopropylacrylamide-co-acrylic acid) (PEP-PEO-P(NIPAm-co-AA)) triblock terpolymers were synthesized by a combination of anionic and RAFT polymerizations, followed by acid hydrolysis. Micellar aggregation and gelation behavior in aqueous solutions were studied by dynamic light scattering (DLS) and rheology, respectively. DLS measurements on dilute solutions revealed that the triblock terpolymers form micelles with PEP cores and PEO-P(NIPAm-co-AA) coronae at room temperature and undergo a micelle to micellar aggregate transition upon heating. Rheological measurements showed that micellar aggregation manifests itself as gelation at higher concentrations (~4 wt %). The observed thermoresponsive aggregation and gelation is due to the intermicellar association of P(NIPAm-co-AA) blocks in the coronae above the lower critical solution temperature of the P(NIPAm-co-AA) block. The critical micellar aggregation and gelation temperatures are controlled by the mole fraction and degree of acrylic acid (AA) ionization in the P(NIPAm-co-AA) block, and therefore they can be modulated as functions of both pH and AA content in the polymer.
Sigma-Aldrich
Poly(N-isopropylacrylamide), carboxylic acid terminated, average Mn 10,000
Sigma-Aldrich
Zinc acrylate, 98%
Sigma-Aldrich
Poly(N-isopropylacrylamide), Mn ~40,000
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Poly(N-isopropyl acrylamide), NHS ester end functionalized, average Mn 5,000