Merck
  • Structure-activity relationship study on benzoic acid part of diphenylamine-based retinoids.
Structure-activity relationship study on benzoic acid part of diphenylamine-based retinoids.
Bioorganic & medicinal chemistry letters (2012-12-12)
Kiminori Ohta, Emiko Kawachi, Koichi Shudo, Hiroyuki Kagechika
ABSTRACT
Based on structure-activity relationship studies of the benzoic acid part of diphenylamine-based retinoids, the potent RXR agonist 4 was derivatized to obtain retinoid agonists, synergists, and an antagonist. Cinnamic acid derivatives 5 and phenylpropionic acid derivatives 6 showed retinoid agonistic and synergistic activities, respectively. The difference of the activities is considered to be due to differences in the flexibility of the carboxylic acid-containing substituent on the diphenylamine skeleton. Compound 7, bearing a methyl group at the meta position to the carboxyl group, was an antagonist, dose-dependently inhibiting HL-60 cell differentiation induced by 3.3 × 10(-10)M Am80.
Sigma-Aldrich
Diphenylamine, puriss. p.a., suitable for redox indicator, ACS reagent, reag. Ph. Eur., ≥98% (GC)
Supelco
Diphenylamine, PESTANAL®, analytical standard
Sigma-Aldrich
Diphenylamine, ReagentPlus®, 99%
Sigma-Aldrich
Diphenylamine, ACS reagent, ≥99%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Supelco
Diphenylamine solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
trans-Cinnamic acid, 97%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium benzoate, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Supelco
trans-Cinnamic acid, analytical standard
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland