Merck
  • The potential of gibberellic acid 3 (GA3) and Tween-80 induced phytoremediation of co-contamination of Cd and Benzo[a]pyrene (B[a]P) using Tagetes patula.
The potential of gibberellic acid 3 (GA3) and Tween-80 induced phytoremediation of co-contamination of Cd and Benzo[a]pyrene (B[a]P) using Tagetes patula.
Journal of environmental management (2012-12-12)
Yuebing Sun, Yingming Xu, Qixing Zhou, Lin Wang, Dasong Lin, Xuefeng Liang
ABSTRACT
The present study was conducted to investigate the effectiveness of GA(3) and Tween-80 on enhancing the phytoremediation of Cd-B[a]P co-contaminated soils. Results showed that the addition of GA(3) and GA(3)-Tween-80 enhanced Tagetes patula growth by 14%-32% and 23%-55%, respectively, relative to the control group. However, under independent GA(3)-treated soils, Cd and B[a]P concentrations in the shoots of the plants decreased by 15%-33% and 15%-53%, respectively, compared with CK. By contrast, the shoot concentration and accumulation of Cd under GA(3)-Tween-80 treatment increased by 0.01-0.46 and 1.33-1.55 times, respectively, whereas those of B[a]P increased from 0.57 to 0.82, and 1.33 to 1.55 times, respectively, compared with those of the control. Optimal result for Cd phytoextraction was obtained under combined 5 mmol Tween-80 kg(-1) and 1 mmol GA(3) kg(-1) treatment, and the maximum removal rate of B[a]P was obtained after the application of 5 mmol Tween-80 kg(-1) and 5 mmol GA(3) kg(-1).
Sigma-Aldrich
Gibberellic acid, 90% gibberellin A3 basis (HPLC)
Sigma-Aldrich
Gibberellic acid, BioReagent, suitable for plant cell culture, ≥90% gibberellin A3 basis (of total gibberellins.)
Supelco
Gibberellic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Gibberellic acid potassium salt, suitable for plant cell culture, BioReagent, ~95%, ≥50% total GA3 basis