Merck
  • Hazardous dyes removal from aqueous solution over mesoporous aluminophosphate with textural porosity by adsorption.
Hazardous dyes removal from aqueous solution over mesoporous aluminophosphate with textural porosity by adsorption.
Journal of hazardous materials (2012-12-19)
Chellapandian Kannan, Kumarasamy Muthuraja, Murugan R Devi
ABSTRACT
Dye pollution in aquatic nature produce serious environmental effects. In this investigation, mesoporous aluminophosphate molecular sieve synthesized and applied for the removal of hazardous dyes Malachite green (MG) and Methylene blue (MB). In the synthesis of mesoporous aluminophosphate (AlPO(4)) molecular sieve, the structure-directing agent, long-chain alkylbenzene has been used as a template. The template used for the synthesis of mesoporous material is environmentally biodegradable. The mesoporous AlPO(4) was synthesized by the absence of an organic base, tetramethyl ammonium hydroxide (TMAOH) which is necessary to maintain the pH for the conventional AlPO(4) synthesis methods. The synthesized mesoporous AlPO(4) has high thermal stability up to 1173K and large porosity nature (40 nm). It was confirmed by the characterization techniques such as low-angle XRD, FT-IR, TGA and BET surface area analysis. The morphology of the material was explained by using SEM and TEM. The hazardous dyes MG and MB removal studied under the various conditions like contact time, dye concentration, temperature, pH and adsorbent dosage to examine the adsorption characteristics of the newly synthesized mesoporous AlPO(4) molecular sieves.
Product Description
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard
Sigma-Aldrich
Aluminum phosphate, reagent grade
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, suitable for microscopy
Sigma-Aldrich
Malachite Green oxalate salt, suitable for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Malachite Green oxalate salt, certified by the BSC
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade
Sigma-Aldrich
Aluminum phosphate monobasic, purum, ≥95.0% (gravimetric)
Sigma-Aldrich
Aluminum phosphate monobasic solution, ~50% in H2O
Sigma-Aldrich
Malachite Green solution, suitable for microscopy