Merck
  • The selective serotonin reuptake inhibitor paroxetine decreases breakpoint of rats engaging in a progressive ratio licking task for sucrose and quinine solutions.
The selective serotonin reuptake inhibitor paroxetine decreases breakpoint of rats engaging in a progressive ratio licking task for sucrose and quinine solutions.
Chemical senses (2012-12-21)
Clare M Mathes, Jillian R Gregson, Alan C Spector
ABSTRACT
Increased serotonergic activity has been shown to reduce motivation to ingest, which may involve, in part, gustatory processes. Here, we examined the effect of paroxetine, a selective serotonin reuptake inhibitor, on appetitive responding for a preferred and an avoided taste solution using a progressive ratio (PR) task in which licking was employed as the operant. Male Sprague-Dawley rats (n = 8/taste stimulus) were trained to respond for a concentration series of sucrose or quinine on fixed and PR schedules of reinforcement. Performance for sucrose was assessed while the rats were partially food- and water-restricted and nondeprived, and performance for water and quinine was assessed while the rats were water-deprived. Then, the rats were injected with vehicle (10% dimethyl sulfoxide, 1mL/kg intraperitoneal [ip], -1h) or paroxetine (5mg/kg), and their responding on a PR schedule for sucrose measured when the rats were nondeprived or for water and quinine when the rats were water-deprived. Paroxetine decreased breakpoint, which was defined as the number of operant (e.g., dry) licks in the final reinforced ratio, for water, quinine, and sucrose. This demonstrates that a general systemic increase in serotonergic activity decreases the appetitive-based responses to both preferred and nonpreferred fluids under different deprivation states.
Product Description
Paroxetine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid
Sigma-Aldrich
Paroxetine hydrochloride hemihydrate, ≥98% (HPLC), powder
Paroxetine hydrochloride hemihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Quinine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Paroxetine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Quinine, 90%