Merck
  • The monolithic lawn-like CuO-based nanorods array used for diesel soot combustion under gravitational contact mode.
The monolithic lawn-like CuO-based nanorods array used for diesel soot combustion under gravitational contact mode.
Nanoscale (2012-12-21)
Yifu Yu, Ming Meng, Fangfang Dai
ABSTRACT
A simple and feasible contact mode called gravitational contact mode (GCM) was developed for the first time to imitate the practical state between soot and catalyst. By simulating rainwater adsorption on a lawn in nature, we synthesized a lawn-like CuO nanorods array, which exhibited rather good catalytic activity for diesel soot combustion under GCM. Moreover, the CuO nanorods array could serve as a support for composite catalysts through a sequential chemical bath deposition method and exhibited higher catalytic activity than a traditional supported catalyst. The monolithic macroscopic structure of such a catalyst shows its potential for large-scale preparation and application.
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%
Sigma-Aldrich
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%