Merck
  • Syntheses, characterization, and optical properties of ternary Ba-Sn-S system compounds: acentric Ba7Sn5S15, centric BaSn2S5, and centric Ba6Sn7S20.
Syntheses, characterization, and optical properties of ternary Ba-Sn-S system compounds: acentric Ba7Sn5S15, centric BaSn2S5, and centric Ba6Sn7S20.
Inorganic chemistry (2012-12-22)
Zhong-Zhen Luo, Chen-Sheng Lin, Wen-Dan Cheng, Hao Zhang, Wei-Long Zhang, Zhang-Zhen He
ABSTRACT
Three new ternary Ba-Sn-S system compounds, acentric Ba(7)Sn(5)S(15), centric BaSn(2)S(5), and centric Ba(6)Sn(7)S(20) have been designed and synthesized by a conventional high-temperature solid-state reaction method using the evacuated silica tubes. The crystal structure of Ba(7)Sn(5)S(15) shows the coexistence of a SnS(4) tetrahedral and a Sn(2)S(3) trigonal bipyramid. Importantly, the larger dipole moment of the [Sn(2)S(3)](2-) trigonal bipyramid group and the polarity enhancement of the bipyramidal arrangements result in a strong SHG effect at 2.05 μm, which is 10 times of the SHG intensity of the benchmark AgGaS(2) with the particle size of 30-46 μm and twice as much as that with the particle size of 150-212 μm. Evidently, the acentric Ba(7)Sn(5)S(15) is a novel IR NLO crystal material with a wide mid-IR window and a strong SHG effect, which is the first reported among the Ba-Sn-S ternary system. Moreover, Ba(7)Sn(5)S(15) can achieve type-I phase-matching that can be used for practical applications. In the centric BaSn(2)S(5,) all Sn atoms are coordinated by five S atoms to form novel SnS(5) trigonal bipyramid polyhedrons. In the other centric Ba(6)Sn(7)S(20), there is the coexistence of the two coordination patterns with a SnS(5) trigonal bipyramid and SnS(4) tetrahedral polyhedrons, featuring a special crystal structure in the Ba-Sn-S system.
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Barium, pieces, 1 cm, 99% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Barium, rod, diam. ~2 cm, ≥99% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Barium, beads, 0.5-2.0 mm particle size, 99% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis