Merck
  • Correlation between emission property and concentration of Sn2+ center in the SnO-ZnO-P2O5 glass.
Correlation between emission property and concentration of Sn2+ center in the SnO-ZnO-P2O5 glass.
Optics express (2012-12-25)
Hirokazu Masai, Toshiro Tanimoto, Takumi Fujiwara, Syuji Matsumoto, Yomei Tokuda, Toshinobu Yoko
ABSTRACT
The authors report on the correlation between the photoluminescence (PL) property and the SnO amount in SnO-ZnO-P2O5 (SZP) glass. In the PL excitation (PLE) spectra of the SZP glass containing Sn2+ emission center, two S1 states, one of which is strongly affected by SnO amount, are assumed to exist. The PLE band closely correlates with the optical band edge originating from Sn2+ species, and they both largely red-shifts with increasing amount of SnO. The emission decay time of the SZP glass decreased with increasing amount of SnO and the internal quantum efficiencies of the SZP glasses containing 1~5 mol% of SnO are comparable to that of MgWO4. It is expected that the composition-dependent S1 state (the lower energy excitation band) governs the quantum efficiency of the SZP glasses.
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin(IV) oxide, nanopowder, ≤100 nm avg. part. size
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, ≥99.99% trace metals basis