Merck
  • Genipin crosslinked ethyl cellulose-chitosan complex microspheres for anti-tuberculosis delivery.
Genipin crosslinked ethyl cellulose-chitosan complex microspheres for anti-tuberculosis delivery.
Colloids and surfaces. B, Biointerfaces (2012-12-26)
Hanzhou Feng, Limei Zhang, Chunyan Zhu
ABSTRACT
Genipin-crosslinked complex microspheres made of the combination of two polymers, ethyl cellulose and chitosan, were prepared by spray drying method. Rifabutin, an anti-tuberculosis agent was used as a model drug. Effects of various specifications of ethyl cellulose and chitosan, different drug/polymers ratios and crosslinking effect of genipin on complex microspheres and drug release characteristics were compared to obtain optimized manufacturing parameters. The complex microspheres showed a significant different shape as compared to chitosan microspheres. Biphasic release features were observed in the in vitro and in vivo release study, consisting of an initial quick release phase and an extended sustained release phase. Furthermore, pulmonary drug concentrations of rats after administering the complex microspheres were maintained on a therapeutic level for at least 24 days.
Sigma-Aldrich
Genipin, ≥98% (HPLC), powder
Sigma-Aldrich
Rifabutin, >98% (HPLC), powder
Sigma-Aldrich
Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
Sigma-Aldrich
Ethyl cellulose, viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis