Merck
  • Naphthalene-based environmentally sensitive fluorescent 8-substituted 2'-deoxyadenosines: application to DNA detection.
Naphthalene-based environmentally sensitive fluorescent 8-substituted 2'-deoxyadenosines: application to DNA detection.
Bioorganic & medicinal chemistry letters (2013-01-01)
Azusa Suzuki, Naoya Takahashi, Yuji Okada, Isao Saito, Nobukatsu Nemoto, Yoshio Saito
ABSTRACT
We synthesized various C8-naphthylethynylated 2'-deoxyadenosine derivatives and investigated their photophysical properties. Among them, cyano- and N,N-dimethylamino-substituted 8-naphthylethynylated 2'-deoxyadenosine derivatives ((cn)A and (dn)A) showed strong fluorescence with high quantum yields and a remarkable solvatofuorochromicity. In particular, fluorescence of N,N-dimethylamino-substituted (2,6dn)A was not quenched by neighboring guanines (Gs) when incorporated in DNA duplexes, in contrast to (cn)A. We developed a new fluorescent probe containing (2,6dn)A that can be used for the detection of target DNA via a bulge formation regardless of the neighboring sequences.
Product Description
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, analytical standard
Supelco
Melting point standard 79-81°C, analytical standard