Merck
  • Fullerene crystals with bimodal pore architectures consisting of macropores and mesopores.
Fullerene crystals with bimodal pore architectures consisting of macropores and mesopores.
Journal of the American Chemical Society (2013-01-02)
Lok Kumar Shrestha, Yusuke Yamauchi, Jonathan P Hill, Kun'ichi Miyazawa, Katsuhiko Ariga
ABSTRACT
A new class of fullerene (C(60)) crystals with bimodal pore architectures consisting of macropores and mesopores was synthesized by using a liquid-liquid interfacial precipitation (LLIP) method involving an interface between isopropyl alcohol (IPA) and a saturated solution of C(60) in a mixture of benzene and carbon tetrachloride (CCl(4)). By varying the mixing fraction of CCl(4) in benzene, the porosity and electrochemically active surface area can be flexibly controlled.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Isopropanol, 70% in H2O