Merck
  • Leptin plasma concentrations increase during antidepressant treatment with amitriptyline and mirtazapine, but not paroxetine and venlafaxine: leptin resistance mediated by antihistaminergic activity?
Leptin plasma concentrations increase during antidepressant treatment with amitriptyline and mirtazapine, but not paroxetine and venlafaxine: leptin resistance mediated by antihistaminergic activity?
Journal of clinical psychopharmacology (2013-01-02)
Claudia Schilling, Maria Gilles, Werner F Blum, Emmerich Daseking, Michael Colla, Bettina Weber-Hamann, Florian Lederbogen, Bertram Krumm, Isabella Heuser, Stefan A Wudy, Daniel Kopf, Michael Deuschle
ABSTRACT
Treatment with several psychopharmacological agents has been associated with increased leptin plasma concentrations. We measured leptin plasma concentrations in 76 adult depressed patients after a 6-day washout phase and again after 35 days of treatment with amitriptyline or paroxetine, as well as in 73 depressed patients after 28 days of treatment with either mirtazapine or venlafaxine. Leptin plasma concentrations increased during treatment with amitriptyline and mirtazapine, even after controlling for increased body mass index and irrespective of response to treatment [14.5 (13.8) vs 20.3 (18.7) ng/mL, and 12.2 (15.8) vs 14.4 (16.5) ng/mL in the 2 cohorts, respectively]. In contrast, paroxetine and venlafaxine treatment was not associated with changes in leptin plasma concentrations [14.8 (12.0) vs 13.6 (10.6); 15.9 (17.3) vs 13.5 (14.6) ng/mL] nor with weight gain. We conclude that treatment with amitriptyline or mirtazapine is associated with an increase in leptin secretion beyond change in weight. Thus, high leptin levels apparently are ineffective in the control of weight gain, indicating leptin resistance. Leptin resistance may be mediated by an antihistaminergic effect on hypothalamic nuclei integrating signals relevant for energy balance.
Product Description
Paroxetine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Venlafaxine hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid
Supelco
Venlafaxine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Mirtazapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Paroxetine hydrochloride hemihydrate, ≥98% (HPLC), powder
Paroxetine hydrochloride hemihydrate, European Pharmacopoeia (EP) Reference Standard
Venlafaxine for system suitability, European Pharmacopoeia (EP) Reference Standard
Venlafaxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Paroxetine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Mianserin hydrochloride