Merck
  • Inter- and intra-specific variability in isoprene production and photosynthesis of Central European oak species.
Inter- and intra-specific variability in isoprene production and photosynthesis of Central European oak species.
Plant biology (Stuttgart, Germany) (2013-01-11)
R Steinbrecher, N Contran, F Gugerli, J-P Schnitzler, I Zimmer, T Menard, M S Günthardt-Goerg
ABSTRACT
European deciduous oaks are closely related and are known for their strong emission of volatile isoprenoids. They are chemo-taxonomically diverse, but hybridise frequently. Four-year-old oak seedlings growing together in a model ecosystem facility under near-natural conditions were studied. The leaves were morphologically classified in the three oak species Quercus robur, Q. pubescens and Q. petraea (with four provenances each) and further investigated by a molecular-genetic approach. Q. robur was morphologically and genetically clearly different from Q. pubescens and Q. petraea, whereas Q. pubescens and Q. petraea individuals used in this study were morphologically and genetically more similar. There was a minor impact of among and within species variability on isoprene synthesis, isoprene emission and photosynthesis. Isoprene emission rates normalised to 25 °C leaf temperature ranged from 5.78 to 10.66 nmol m(-2)  s(-1) , whereas photosynthesis ranged from 12.8 to 17.6 μmol m(-2)  s(-1) . On cloudy days, among the provenances of each species, only net photosynthesis of the Q. robur provenance Hünenberg was reduced and isoprene synthase activity of the Q. pubescens provenance Promotogno increased. On sunny days, photosynthesis did not differ among the provenances. Over all provenances, gas exchange on cloudy days did not differ significantly from sunny days. In the combined data of cloudy and sunny days, no differences between the studied provenances and oak species were detected in isoprene emission and photosynthesis. Thus, isoprene emission and photosynthesis rates were remarkably stable among oak species and provenances. The results indicate that taxonomic differences in the studied oak species are not reflected in isoprene emission and photosynthesis, probably because of the high plasticity of gene expression resulting in high phenotypic flexibility.
Product Description
Supelco
Isoprene, analytical standard
SAFC
Isoprene, ≥99%
Sigma-Aldrich
Isoprene, 99%, contains <1000 ppm p-tert-butylcatechol as inhibitor