Merck
  • [Influence of exogenous embryonic fibroblasts on collagen type I and type III ratio in the tissues of paraprosthetic capsule (experimental work)].
[Influence of exogenous embryonic fibroblasts on collagen type I and type III ratio in the tissues of paraprosthetic capsule (experimental work)].
Tsitologiia (2013-01-05)
I S ivanov, V A Lazarenko, S V Ivanov, G N Goriainova, A V Ivanov
ABSTRACT
Nowadays, the use of synthetic prosthesis is obligatory method in surgical treatment of large and giant ventral hernias. We have fulfilled comparative investigation of the dynamics of collagen Type I to Type III ratio in paraprosthetic region in mice when using implants, Esphyl - polyethylene, Ecophlon - polytetrafluorethylene, and Uniflex - polyvinylidenfluoride, with a one- and two-time introduction of cultured fibroblasts into paraprosthetic region at different times after implantations and without the introduction. Use of Sirius Red dye staining and polarization microscopy revealed that, in short period (10 days after endoprosthesis implantation), the implication of fibroblasts and the number of implications did not affect the collagen Types ratio. In later stages (30-60 days after endoprosthesis implantation) a significant increase in collagen Type I was observed when using all the materials. The ratio of collagen Type I to Type III was maximal at all stages of the experiment in the case of the implant Uniflex. Double implication of exogenous fibroblasts accelerated the increase in the collagen Type I to Type III ratio to a greater extent.
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~530,000, pellets
Sigma-Aldrich
Poly(vinylidene fluoride), powder
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~180,000 by GPC, average Mn ~71,000, beads or pellets
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~275,000 by GPC, average Mn ~107,000, pellets