Merck
  • Determination of malachite green, crystal violet and their leuco-metabolites in fish by HPLC-VIS detection after immunoaffinity column clean-up.
Determination of malachite green, crystal violet and their leuco-metabolites in fish by HPLC-VIS detection after immunoaffinity column clean-up.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-01-05)
Jie Xie, Tao Peng, Dong-Dong Chen, Qing-Jie Zhang, Guo-Min Wang, Xiong Wang, Qi Guo, Fan Jiang, Dan Chen, Jian Deng
ABSTRACT
A high performance liquid chromatography method with visible detection (HPLC-VIS) for the determination of malachite green (MG), crystal violet (CV), leucomalachite green (LMG), and leucocrystal violet (LCV) in fish has been developed after clean-up through an immunoaffinity column (IAC). Residues were simultaneously extracted from fish muscle with acetonitrile and ammonium acetate buffer. The leuco-forms, LMG and LCV, were oxidized quantitatively to the chromic CV and MG by reaction with 2,3-dichloro-5,6-dicyano-1,4-benzoquinone. Extracts were then purified on an IAC which prepared by immobilizing the anti-MG-CV antibodies by the sol-gel method. Finally, the eluents were analyzed by HPLC-VIS. The limits of detection were 0.15, 0.1, 0.18 and 0.14ng/g for MG, CV, LMG and LCV, respectively. The average recoveries in samples fortified with MG, CV, LMG and LCV over the range 0.5-10ng/g were from 71.6% to 96.8% with RSDs of 5.1-12.3% (n=6). This novel method was confirmed by liquid chromatography-tandem mass spectrometry with electrospray interface in positive mode using multiple reaction monitoring.
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution A, suitable for microscopy
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade
Sigma-Aldrich
Malachite Green oxalate salt, certified by the BSC
Sigma-Aldrich
Malachite Green oxalate salt, suitable for microscopy, crystalline, S. No.: 754
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard
Sigma-Aldrich
Malachite Green solution, suitable for microscopy
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
Leucomalachite Green, powder
Sigma-Aldrich
Crystal Violet, suitable for microscopy
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet Solution
Supelco
Leucomalachite Green, analytical standard
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis