Merck
  • Burdock fermented by Aspergillus awamori elevates cecal Bifidobacterium, and reduces fecal deoxycholic acid and adipose tissue weight in rats fed a high-fat diet.
Burdock fermented by Aspergillus awamori elevates cecal Bifidobacterium, and reduces fecal deoxycholic acid and adipose tissue weight in rats fed a high-fat diet.
Bioscience, biotechnology, and biochemistry (2013-01-08)
Yukako Okazaki, Novita Vivi Sitanggang, Satoko Sato, Nanae Ohnishi, Junji Inoue, Takafumi Iguchi, Toshiro Watanabe, Hiroyuki Tomotake, Kazuki Harada, Norihisa Kato
ABSTRACT
This study investigated the effects of dietary supplementation with burdock powder and Aspergillus awamori-fermented burdock powder at 5% on the intestinal luminal environment and body fat in rats fed a high-fat (HF) diet. Food intake and growth were unaffected by dietary manipulation. Consumption of the burdock and fermented burdock diets significantly elevated fecal IgA and mucins (indices of intestinal immune and barrier functions) and reduced fecal lithocholic acid (a risk factor for colon cancer) (p<0.05). The fermented burdock diet markedly elevated cecal Bifidobacterium and organic acids, including lactate, acetate, propionate, and butyrate, and reduced fecal deoxycholic acid (a risk factor for colon cancer) and perirenal adipose tissue weight (p<0.05), but the burdock diet did not. These results suggest that consumption of fermented burdock improves the intestinal luminal environment and suppresses obesity in rats fed a HF diet.
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Butyric acid, ≥99%, FG
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%
Sigma-Aldrich
Propionic acid, ≥99.5%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)