Merck
  • In vitro plant propagation of Catharanthus roseus and assessment of genetic fidelity of micropropagated plants by RAPD marker assay.
In vitro plant propagation of Catharanthus roseus and assessment of genetic fidelity of micropropagated plants by RAPD marker assay.
Applied biochemistry and biotechnology (2013-01-08)
Ajay Kumar, Krishna Prakash, Rajesh Kumar Sinha, Nitish Kumar
ABSTRACT
An investigation was carried out to develop an efficient micropropagation protocol for Catharanthus roseus. Experiments were conducted to optimize suitable media for in vitro shoot multiplication and root induction. Out of the different media compared for in vitro shoot multiplication, Murashige and Skoog (MS) medium supplemented with 1 mg/l of 6-benzylaminopurine and 0.2 mg/l α-naphthaleneacetic acid showed better response in terms of the emergence of shoots from axillary buds as well as proliferation and multiplication of shoots. The shoots when placed on half strength of MS medium having 1 mg/l indole 3-butyric acid and 0.25 % charcoal showed cent percent root induction with maximum number of roots per shoot (4.2) as well as maximum root length (1.72 cm). Further, clonal fidelity of the in vitro-raised plants was carried out using randomly amplified polymorphic DNA marker and results indicated that all the tissue culture-derived plants are true-to-type and there were no somaclonal variations among these plants.
Sigma-Aldrich
6-Benzylaminopurine, ReagentPlus®, ≥99.0% (HPLC)
Sigma-Aldrich
6-Benzylaminopurine, suitable for plant cell culture
Sigma-Aldrich
Kinetin, ≥99.0% (HPLC)
Sigma-Aldrich
Kinetin, BioReagent, suitable for plant cell culture, amorphous powder
Sigma-Aldrich
Kinetin, suitable for plant cell culture, crystalline
Supelco
6-Benzylaminopurine, PESTANAL®, analytical standard
Sigma-Aldrich
Kinetin solution, 1 mg/mL, BioReagent, suitable for plant cell culture