Merck
  • Structure of a piperidine-modified calix[4]arene derivative and spectral resolution of its interaction with rare earth metals with chemometric methods.
Structure of a piperidine-modified calix[4]arene derivative and spectral resolution of its interaction with rare earth metals with chemometric methods.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-01-09)
Li Wang, Xiaoya Wang, Yanmei Wang
ABSTRACT
A piperidine-modified calix[4]arene derivative was synthesized and its structure was confirmed with X-ray diffraction data. UV-visible spectroscopy was used to study its molecular recognition of rare earth ions. The results revealed the calix[4]arene derivative could separate tight metal picrate ion pairs by complexation with the rare earth metal ions in tetrahydrofuran. Resolution of the UV-visible spectra with chemometric methods revealed that the derivative and the rare earth ions Eu(3+), Dy(3+), and Tb(3+) formed ML(2) complexes with stability constants of 10(8.26), 10(8.29), and 10(7.41) respectively.
Sigma-Aldrich
Piperidine solution, suitable for peptide synthesis, 20% in DMF
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free