Merck
  • A probe on the intermolecular forces in diisopropyl ether-n-butyric acid mixture by dielectric, FTIR studies and quantum chemical calculations.
A probe on the intermolecular forces in diisopropyl ether-n-butyric acid mixture by dielectric, FTIR studies and quantum chemical calculations.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-01-09)
G Arivazhagan, R Shanmugam, A Elangovan
ABSTRACT
The results of FTIR spectral measurement on equimolar diisopropyl ether-butyric acid binary mixture and quantum chemical calculations on the complex molecule have been presented. Dielectric studies have been carried out on the binary mixture over the entire composition range and at four different temperatures 303 K, 308 K, 313 K and 318 K. n-Butyric acid seems to prefer less polar ether to interact with it. It appears that the usual interpretation of variation of static dielectric constant and positive deviation of excess permittivity from ideal mixture behavior needs to be relooked.
Product Description
Supelco
Diisopropyl ether, analytical standard
Sigma-Aldrich
Diisopropyl ether, anhydrous, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Diisopropyl ether, ReagentPlus®, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Diisopropyl ether, contains either BHT or hydroquinone as stabilizer, ACS reagent, ≥99.0%
Sigma-Aldrich
Diisopropyl ether, contains either BHT or hydroquinone as stabilizer, ACS reagent, ≥99.0%
Sigma-Aldrich
Diisopropyl ether, puriss. p.a., ≥98.5% (GC)
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Butyric acid, ≥99%
Supelco
Butyric acid, analytical standard