Merck
  • Synthesis, characterization, and biological evaluation of new tetrazole-based platinum(II) and palladium(II) chlorido complexes--potent cisplatin analogues and their trans isomers.
Synthesis, characterization, and biological evaluation of new tetrazole-based platinum(II) and palladium(II) chlorido complexes--potent cisplatin analogues and their trans isomers.
Journal of inorganic biochemistry (2013-01-12)
Tatiyana V Serebryanskaya, Tatiana Yung, Alexey A Bogdanov, Andrei Shchebet, Steven A Johnsen, Alexander S Lyakhov, Ludmila S Ivashkevich, Zhanna A Ibrahimava, Tatiyana S Garbuzenco, Tatiyana S Kolesnikova, Natalya I Melnova, Pavel N Gaponik, Oleg A Ivashkevich
ABSTRACT
Two series of tetrazole-containing platinum(II) and palladium(II) chlorido complexes, trans-[ML(2)Cl(2)] (M=Pt, Pd) and cis-[PtL(2)Cl(2)]·nH(2)O (n=0, 1), where L is 1- or 2-substituted 5-aminotetrazole, have been synthesized and thoroughly characterized. Configuration of platinum(II) complexes obtained from the reaction of 5-aminotetrazoles with K(2)PtCl(4) has been found to vary depending on the nature of tetrazole derivatives and reaction conditions. According to in vitro cytotoxic evaluation, only platinum complexes display noticeable antiproliferative effect, and their cytotoxicity depends strongly on their geometry and hydrophobicity of the carrier ligands. The most promising complexes are cis-[Pt(1-apt)(2)Cl(2)]·H(2)O and cis-[Pt(2-abt)(2)Cl(2)]·H(2)O, where 1-apt is 5-amino-1-phenyltetrazole and 2-abt is 5-amino-2-tert-butyltetrazole. In comparison with cisplatin, they show comparable cytotoxic potency against cisplatin-sensitive human cancer cell lines, cis-[Pt(2-abt)(2)Cl(2)]·H(2)O performing substantially higher activity against cisplatin-resistant cell lines. Cell cycle studies in H1299 cell line indicated that cis-[Pt(2-abt)(2)Cl(2)]·H(2)O induced apoptosis launched from G2 accumulations. The DNA interaction with cis-[Pt(1-apt)(2)Cl(2)]·H(2)O was followed by UV spectroscopy, circular dichroism, hydrodynamic and electrophoretic mobility studies. Both cis-[Pt(1-apt)(2)Cl(2)]·H(2)O and cis-[Pt(2-abt)(2)Cl(2)]·H(2)O complexes appeared to be significantly less toxic than cisplatin in mice, while only compound cis-[Pt(1-apt)(2)Cl(2)]·H(2)O displayed noticeable efficacy in vivo.
Sigma-Aldrich
5-Aminotetrazole, 97%
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Type XV, Activated, lyophilized powder
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Genomic, unsheared
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from calf thymus, lyophilized powder